Recombinant dengue NS1 ST1
ENZOENZPRT1040100
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Seq:02
- Protein synonyms:non-structural protein|viral|strain Nauru/Western Pacific/1974|DENV1-NS1
- Protein/peptide name:Dengue NS1 ST1
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid, In DPBS pH 7,4
- Purification:Purified protein
Dengue virus NS1 protein serotype 1 presented in its native folded state with all post-translational modifications to deliver optimal antigenicity.
Produced in HEK 293 cells. Dengue Viral NS1 Serotype 1 (DENV1-NS1) from strain Nauru/Western Pacific/1974.