Order Entry
Belgium
ContactUsLinkComponent
Human recombinant FGF-basic 154
Human recombinant FGF-basic 154
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant FGF-basic 154
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Biological activity:
    The activity is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells which is typically observed at concentations <1 ng/ml.
  • Protein synonyms:
    Prostatropin|HBGF-2|FGF2
  • Protein/peptide name:
    FGF-basic 154
  • Purity:
    >97% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
  • Molecular weight:
    ~17,2kDa
  • Formulation:
    Lyophilized from 10 mM sodium phosphate, 75 mM NaCl, pH 7,5. Sterile filtered.

Specifications

Frequently Bought Together