Human recombinant IL-3
Catalog # ENZOALX2018200010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:The activity is determined by the dose-dependent stimulation of TF-1 cells which is typically observed at concentations <0,1 ng/ml.
- Protein synonyms:MCGF|HCGF|Multi-CSF|P-cell stimulation factor|Interleukin-3
- Protein/peptide name:IL-3
- Purity:>95% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- Molecular weight:~15kDa
- Formulation:Lyophilized. Sterile filtered.
Specifications
About this item
Produced in E. coli. Non-glycosylated protein, containing 133 amino acids.
- Carrier-free