Human recombinant IL-6
Catalog # ENZOALX2018180020
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Biological activity:The activity is determined by the dose-dependant stimulation of mouse 7TD1 cells which is typically observed at concentrations <1 ng/ml.
- Protein synonyms:Interleukin-6|HSF|BSF-2|IFN-beta2|B-cell differentiation factor|MGI-2
- Protein/peptide name:IL-6
- Purity:>95% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- Molecular weight:~21kDa
- Formulation:Lyophilized from 10 mM acetic acid. Sterile filtered.
Specifications
About this item
Produced in E. coli. Non-glycosylated protein, containing 184 amino acids.
- Carrier-free