Order Entry
Belgium
ContactUsLinkComponent
 
Rat recombinant epidermal growth factor
Catalog # ENZOALX2018120100
Supplier:  Enzo Life Sciences
CAS Number:  
Rat recombinant epidermal growth factor
Catalog # ENZOALX2018120100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Seq:
    02
  • Biological activity:
    The activity is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells which is typically observed at concentations <0,1 ng/ml.
  • Protein synonyms:
    Urogastrone|EGF|URG
  • Protein/peptide name:
    Epidermal Growth Factor
  • Purity:
    >95% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
  • Molecular weight:
    ~6,3kDa
  • Formulation:
    Lyophilized from 10 mM Na2PO4, pH 7,5. Sterile filtered.

Specifications

About this item

Produced in E. coli. Non-glycosylated protein, containing 54 amino acids, with a molecular weight of 6.3 kDa.

  • Carrier-free