Human recombinant myeloperoxidase
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Isolated from human leukocytes. Negative for HBsAG-, HCV- and HIV-antibodies.££Haem protein that catalyzes oxidations by H₂O₂, including MPO-chloride-mediated killing of microbes and tumor cells, inactivation of chemotactic factors, cross-linking of proteins and iodination of proteins.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:MPO
- Protein/peptide name:Myeloperoxidase
- Purity:>98%
- Formulation:Lyophilised from 5 mM phosphate buffer, pH 7,4.
Specifications
Frequently Bought Together