Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant Erp72,HIS Tag
 
Human recombinant Erp72,HIS Tag

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    Erp72
  • Purity:
  • Formulation:
    Liquid
  • Purification:
  • Molecular weight:
    ~68 kDa

 

Frequently Bought Together