Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Hsp 60
Human recombinant Hsp 60
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant Hsp 60
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    Hsp 60
  • Purity:
  • Formulation:
    Liquid
  • Purification:
  • Molecular weight:
    ~60 kDa

Specifications

Frequently Bought Together