Human recombinant alpha-A-Crystallin,HIS tag
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli. Human αA-Crystallin is fused at the N-terminus to a His-tag.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Tag sequence:HIS Tag
- Protein synonyms:HSPB4|CRYAA|HspB4|Short Form|Heat Shock Protein Beta-4|Alpha-Crystallin A Chain|CRYA1
- Protein/peptide name:alpha-A-Crystallin
- Purity:-
- Formulation:Liquid
- Purification:-
- Molecular weight:~20 kDa
Specifications
Frequently Bought Together