Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant DKK1,HIS Tag
Mouse recombinant DKK1,HIS Tag
Fournisseur:  ENZO LIFE SCIENCES
Certificats
Mouse recombinant DKK1,HIS Tag
Fournisseur:  ENZO LIFE SCIENCES

Spécifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    DKK1
  • Purity:
    >95% (SDS-PAGE; Western blot)
  • Molecular weight:
    ~35 kDa
  • Formulation:
    Liquid, In 50 mM TRIS, pH 8,5, containing 250 mM sodium chloride, 2 mM DTT, and 1 mM EDTA
  • Purification:
    Purified by multi-step chromatography

Spécifications

Frequently Bought Together