Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Lc3-I,HIS Tag
Human recombinant Lc3-I,HIS Tag
Supplier:  ENZO LIFE SCIENCES
Certificates
Human recombinant Lc3-I,HIS Tag
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    Lc3-I
  • Purity:
    >95% (SDS-PAGE)
  • Molecular weight:
    ~16 kDa (predicted), 20 kDa (observed)
  • Formulation:
    Liquid. In 25 mm TRIS, pH 7,5, containing 250 mm sodium chloride, 1,0 mm EDTA, and 1,0 mm DTT
  • Purification:
    Purified by multi-step chromatography

Specifications

Frequently Bought Together