Order Entry
Belgium
ContactUsLinkComponent
Native Hsv-2 antigen
Native Hsv-2 antigen
Catalog # ENZOALX6301270200
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Native Hsv-2 antigen
Catalog # ENZOALX6301270200
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    200 µl
  • Protein/peptide type:
    Native
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Protein/peptide name:
    Hsv-2 Antigen
  • Purity:
    ~80%
  • Formulation:
    Liquid, PBS containing 0,1% Triton X-100

Specifications

About this item

From herpes simplex virus type 2, strain G. Inactivated by 0,1% Triton X-100.