Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant USP14 (from
E. coli
)
Human recombinant USP14 (from E. coli)
ENZOBMLUW98400100
:
ENZO LIFE SCIENCES
:
Human recombinant USP14 (from E. coli)
ENZOBMLUW98400100
:
ENZO LIFE SCIENCES
:
Pk:
100 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Seq:
02
Protein/peptide name:
USP14
Produced in
E. coli
.