Human recombinant UbcH13/Mms2 (from E. coli), HIS tag
Catalog # ENZOBMLUW95650100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein synonyms:Mms2
- Protein/peptide name:UbcH13/Mms2
- Purity:>95% (SDS-PAGE)
- Concentration:1,0 mg/ml
- Formulation:Liquid. In 50 mM HEPES, pH 8,0, containing 50 mM sodium chloride, 10% Glycerol, and 1 mM DTT.
Specifications
About this item
Produced in E. coli.
A heterodimer composed of the catalytically active ubiquitin-conjugating enzyme UbcH13 and its catalytically inactive paralog hMms2, which lacks an active site cysteine residue.