Order Entry
Belgium
ContactUsLinkComponent
Nona-Ubiquitin (linear) (from E. coli)
Nona-Ubiquitin (linear) (from E. coli)
Catalog # ENZOBMLUW08100100
Supplier:  Enzo Life Sciences
CAS Number:  
Nona-Ubiquitin (linear) (from E. coli)
Catalog # ENZOBMLUW08100100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Source:
    E. coli
  • Seq:
    02
  • Protein synonyms:
    UB9
  • Protein/peptide name:
    Nona-Ubiquitin (linear)
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    1 mg/ml
  • Formulation:
    Liquid. In 50 mM TRIS, pH 8,0, containing 50 mM sodium chloride.

Specifications

About this item

Produced in E. coli.

Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.