Tetra-ubiquitin (linear) (from Escherichia coli)
ENZOBMLUW07850100
:
- Pk:100 µG
- Source:E. coli
- Seq:02
- Protein synonyms:UB4
- Protein/peptide name:Tetra-Ubiquitin (linear)
- Purity:>95% (SDS-PAGE)
- Concentration:1 mg/ml
- Formulation:Liquid. In 50 mM TRIS, pH 8,0, containing 50 mM sodium chloride.
Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.