Di-Ubiqutin (K63 linked)
Catalog # ENZOBMLUW07300050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Seq:02
- Protein synonyms:UB2
- Protein/peptide name:Di-Ubiqutin (K63 linked)
- Purity:>95% (SDS-PAGE)
- Concentration:2,5 mg/ml
- Formulation:Liquid. In 50 mM TRIS, pH 7,6, containing 1 mM EDTA.
Specifications
About this item
Protein components produced in E. coli. Synthesised enzymatically in vitro.
Polyubiquitin chains, linked through specific lysine residues, are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.