([K48-only]Ub)n-ubiquitinylated Substrate
Catalog # ENZOBMLUW06400025
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:25 µG
- Seq:02
- Protein/peptide name:([K48-only]Ub)n-ubiquitinylated Substrate
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid. In 50mM TRIS, pH 7.5, containing 150mM NaCl and 1mM DTT.
Specifications
About this item
Protein components produced in E. coli. Synthesized enzymatically in vitro. (K48-only)Ubn-ubiquitinylated substrate protein was produced using K48-only mutant ubiquitin.