Human recombinant MMP-19 (catalytic domain)
Catalog # ENZOBMLSE5610010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein/peptide name:MMP-19 (catalytic domain)
- Purity:>95% (SDS-PAGE)
- Concentration:0,5 μg/μl
- Formulation:Liquid. In 25 mM NaBorate, pH 9,3, containing 5 mM CaCl₂, 20 μM ZnCl₂, 0,05% Brij-35 and 20% glycerol.
Specifications
About this item
Produced in E.coli.
Active recombinant MMP-19. MMP-19 is expressed widely, and degrades aggrecan, fibronectin, type I gelatin, and basement membrane components such as laminin, nidogen, and type IV collagen. It is involved in angiogenesis, cancer, rheumatoid arthritis, multiple sclerosis, and psoriasis.