Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant Brain derived neurotrophic factor (from E. coli)
 
Human recombinant Brain derived neurotrophic factor (from E. coli)

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    Brain derived neurotrophic factor
  • Purity:
    >97%

 

Frequently Bought Together