Order Entry
Belgium
ContactUsLinkComponent
Human recombinant LSD1 (from E. coli)
Human recombinant LSD1 (from E. coli)
Catalog # ENZOBMLSE5440050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant LSD1 (from E. coli)
Catalog # ENZOBMLSE5440050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    LSD1
  • Purity:
    >90% (SDS-PAGE)
  • Formulation:
    Liquid. Solution containing 6 mM Na2HPO4, 1,1 mM K2HPO4, pH 7,2, 82 mM sodium chloride, 1,6 mM KCl and 40% (v/v) glycerol.

Specifications

About this item

Highly active

Produced in E. coli.