Order Entry
Belgium
ContactUsLinkComponent
Human recombinant DPP9
Human recombinant DPP9
Catalog # ENZOBMLSE5280010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant DPP9
Catalog # ENZOBMLSE5280010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    DPP9

Specifications

About this item

Produced in insect cells. Active recombinant human DPP9 with an N-terminal purification tag.