Human recombinant PKCalpha (GST-tag),GST tag
ENZOBMLSE4940005
:
- Pk:5 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:PKCalpha (GST-tag)
Produced in insect cells. Full length active PKCa (protein kinase C alpha) is fused at the N-terminus to a GST-tag.