Human Recombinant 14-3-3theta/tau (from E. coli), HIS Tag
Catalog # ENZOBMLSE4870100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:14-3 -3 theta/tau
- Purity:>90% (SDS-PAGE)
- Formulation:Liquid. In 50 mM NaH2PO4 containing 300 mM sodium chloride, pH 8,0.
Specifications
About this item
Produced in E. coli. Human 14-3-3τ/θ is fused at the C-terminus to a His-tag.