Order Entry
Belgium
ContactUsLinkComponent
Human recombinant CDK5/p25 (GST-tag),GST Tag
Human recombinant CDK5/p25 (GST-tag),GST Tag
Catalog # ENZOBMLSE4710005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant CDK5/p25 (GST-tag),GST Tag
Catalog # ENZOBMLSE4710005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    GST Tag
  • Protein/peptide name:
    CDK5/p25 (GST-tag)

Specifications

About this item

Produced in insect cells. Full length active enzyme.