Human recombinant MKP-5 (from insect cells (Sf9)), HIS Tag
ENZOBMLSE4670100
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:MKP-5
Produced in Sf9 cells. Recombinant human MKP-5 fused at the N-terminus to a His-tag.