Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Akt1 (GST-tag)
Human recombinant Akt1 (GST-tag)
Supplier:  Enzo Life Sciences
Certificates
Human recombinant Akt1 (GST-tag)
Supplier:  Enzo Life Sciences

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    GST Tag
  • Protein/peptide name:
    Akt1 (GST-tag)

Specifications

Frequently Bought Together