Order Entry
Belgium
ContactUsLinkComponent
Human recombinant PTEN (from E. coli), HIS Tag
Human recombinant PTEN (from E. coli), HIS Tag
Catalog # ENZOBMLSE4020010
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant PTEN (from E. coli), HIS Tag
Catalog # ENZOBMLSE4020010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    PTEN
  • Purity:
    >85% (SDS-PAGE)
  • Formulation:
    Liquid. In 50 mM TRIS-HCl, pH 8,0, containing 150 mM sodium chloride, 5 mM DTT, 0,03% Brij 35, 0,1 mM EDTA and 20% glycerol.

Specifications

About this item

Produced in E. coli. Full length human PTEN (aa 1-403) is fused at the N-terminus to a His-tag.