Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant Chk1 (GST-tag),GST Tag
  ENZOBMLSE3780005
 :  
Human recombinant Chk1 (GST-tag),GST Tag
  ENZOBMLSE3780005
 :  

 

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    GST Tag
  • Protein/peptide name:
    Chk1 (GST-tag)

 

 

Produced in insect cells. Full length active enzyme.