Human recombinant PKCν (GST-tag),GST tag
Catalog # ENZOBMLSE3490005
Fournisseur: Enzo Life Sciences
Numéro CAS:
Spécifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:PKCν (GST-tag)
Spécifications
À propos de cet article
Produced in insect cells. Active, full length protein kinase C nu (PKCν, protein kinase D3, PKD3) is fused at the N-terminus to a GST-tag.