Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Tau-383 (0N4R variant) (from E. coli)
Human recombinant Tau-383 (0N4R variant) (from E. coli)
Catalog # ENZOBMLSE3240100
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant Tau-383 (0N4R variant) (from E. coli)
Catalog # ENZOBMLSE3240100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Tau-383 (0N4R variant)
  • Formulation:
    Lyophilised

Specifications

About this item

Produced in E. coli.