Human recombinant Tau-441 (2N4R variant) (from E. coli)
Catalog # ENZOBMLSE3210100
Fournisseur: ENZO LIFE SCIENCES
Numéro CAS:
Spécifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Reconstitution instructions:Reconstitute with distilled water to a concentration of 1 mg/ml.
- Protein/peptide name:Tau-441 (2N4R variant)
- Purity:>90% (SDS-PAGE)
- Formulation:Lyophilised from 50 mM MES, pH 6,8, 100 mM sodium chloride and 0,5 mM EGTA.
Spécifications
À propos de cet article
Produced in E. coli.