Human recombinant MMP-11 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE2820010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:MMP-11 (catalytic domain)
Specifications
About this item
Produced in E. coli. Active Matrix Metalloproteinase-11 (MMP-11, Stromelysin-3) catalytic domain from human cDNA.
- Consists of the catalytic domain of human MMP-11 (Phe98-Ser266, NM_005940) with a C-terminal purification tag
MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected. It may be an important link between obesity and cancer.