Order Entry
Belgium
ContactUsLinkComponent
Human recombinant ADAM17 (catalytic domain), HIS-tag
Human recombinant ADAM17 (catalytic domain), HIS-tag
Catalog # ENZOBMLSE2680010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant ADAM17 (catalytic domain), HIS-tag
Catalog # ENZOBMLSE2680010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    ADAM17 (catalytic domain)
  • Purity:
    >90% (SDS-PAGE).
  • Concentration:
    0,45 µg/µl
  • Formulation:
    Liquid. In 22,5 mM TRIS, pH 7,5, containing 4,5µM ZnCl2, 0,0045% Brij-35 and 10% glycerol.

Specifications

About this item

Produced in insect cells.