Human recombinant Akt2 (GST-tag)
ENZOBMLSE2470020
:
- Pk:20 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:Akt2 (GST-tag)
Produced in a baculovirus expression system. The deltaPH domain of human Akt2 (PKBß, Protein Kinase B beta) (aa 119-481) is fused at the N-terminus to a GST-tag.