Saisie de commande
Belgium
ContactUsLinkComponent
 
Human recombinant MMP-13 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE2460010
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant MMP-13 (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE2460010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Cdt:
    10 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Nom de la protéine/du peptide:
    MMP-13 (catalytic domain)

Specifications

About this item

Produced in E. coli. Active Matrix Metalloproteinase-13 (MMP-13, collagenase-3) catalytic domain from human cDNA.

The enzyme consists of the catalytic domain of human MMP-13 (Tyr104-Asn274, NM_002427) with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-13 which lacks the C-terminal hemopexin domain.

MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.