Order Entry
Belgium
ContactUsLinkComponent
Ubiquitin-AMC
Ubiquitin-AMC
Catalog # ENZOBMLSE2110025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Ubiquitin-AMC
Catalog # ENZOBMLSE2110025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Seq:
    02
  • Protein/peptide name:
    Ubiquitin-AMC
  • Purity:
    >95% (HPLC)
  • Formulation:
    In DMSO

Specifications

About this item

High purity industry standard substrate for deubiquitinylating enzymes (DUBs).

Ubiquitin-AMC is a fluorogenic substrate for a wide range of deubiquitinylating enzymes (DUBs), including ubiquitin C-terminal hydrolases (UCHs) and ubiquitin specific proteases (USPs). It is a particularly useful reagent for the study of deubiquitinylating activity where detection sensitivity or continuous monitoring of activity is essential. Ubiquitin-AMC has been shown to be a sensitive substrate for UCH-L3 (Km = 0,039 μM) (Prod. No. BML-UW9745) and for Isopeptidase-T (Km = 0,17-1,4 μM) (Prod. No. BML-UW9690), and is particularly useful for studying deubiquitinylating activity where detection sensitivity or continuous monitoring of activity is essential.