Order Entry
Belgium
ContactUsLinkComponent
Human native cathepsin B (from liver)
Human native cathepsin B (from liver)
  ENZOBMLSE1980025
 :  
Human native cathepsin B (from liver)
  ENZOBMLSE1980025
 :  

 

  • Pk:
    25 µG
  • Protein/peptide type:
    Native
  • Source:
    Liver
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Cathepsin B
  • Grade:
    Purified

 

 

Purified from human liver.