Order Entry
Belgium
ContactUsLinkComponent
Human recombinant retinoid X receptor beta (from insect cells (Sf9))
Human recombinant retinoid X receptor beta (from insect cells (Sf9))
Catalog # ENZOBMLSE1270050
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant retinoid X receptor beta (from insect cells (Sf9))
Catalog # ENZOBMLSE1270050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Retinoid X receptor beta
  • Concentration:
    2 mg/ml total protein (~0,6 mg/ml RXRß)
  • Purification:
    Partially purified.

Specifications

About this item

Produced in Sf9 cells. Full length RXRß (rH2RIIBP).