Human recombinant MMP3 (catalytic domain) (100-272) (from E. coli)
Catalog # ENZOBMLSE1090010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:MMP3 (catalytic domain) (100-272)
Specifications
About this item
Produced in E. coli. Active Matrix Metalloproteinase-3 (MMP-3, stromelysin-1, transin) catalytic domain from human cDNA.
The enzyme consists of the catalytic domain of human MMP-3 (Phe100-Thr272, NM_002422) with a C-terminal purification tag.