pp60v-src
ENZOBMLP3120001
:
- Pk:1 mg
- Seq:02
- Protein/peptide name:pp60v-src
PTP substrate
This sequence, RRLIEDNEpYTARG, is the v-Src (412-422) autophosphorylation site and this peptide contains a phosphorylated tyrosine. Useful in a nonradioactive tyrosine phosphatase assay and as a competitive tyrosine phosphatase inhibitor.