RR-src peptide
ENZOBMLP3080001
New Product
:
- Pk:1 mg
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:RR-src Peptide
- Purity:97%
Src kinase substrate
This tyrosine kinase substrate sequence, RRLIEDAEYAARG, was derived from the v-Src autophosphorylation site (original src sequence is IEDNEYTARQG). Km=5 mM in LSTRA cell extracts.