Order Entry
Belgium
ContactUsLinkComponent
 
AMARA substrate peptide
  ENZOBMLP2700001
 :  
AMARA substrate peptide
  ENZOBMLP2700001
 :  

 

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    AMARA Substrate Peptide
  • Purity:
    >95% (HPLC)
  • Formulation:
    Lyophilized

 

 

AMPK and SIK kinase substrate

This peptide, AMARAASAAALARRR, is a substrate for AMPK and SIK kinases.