Src substrate II
Catalog # ENZOBMLP2590001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:Src Substrate II
- Purity:>95% (HPLC)
- Formulation:Lyophilised
Specifications
About this item
This peptide, YIYGSFK, was derived through a random combinatorial peptide library method and is a specific and efficient substrate for the scr family of kinases. The Km of YIYGSFK for p60c-src is 55 µM.