Order Entry
Belgium
ContactUsLinkComponent
MeOSuc-AAPV-AMC
MeOSuc-AAPV-AMC
Catalog # ENZOBMLP2240005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
MeOSuc-AAPV-AMC
Catalog # ENZOBMLP2240005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    02
  • Protein/peptide name:
    MeOSuc-AAPV-AMC
  • Purity:
    98%

Specifications

About this item

Elastase substrate

Highly sensitive substrate for neutrophil elastase (leukocyte elastase) (Km=130 µM), but not granzyme H. This substrate can be used for in vitro assays with purified enzyme, on live cells, or in conditioned medium. Ex: 380 nm, Em: 460 nm.