Abl kinase substrate
Catalog # ENZOBMLP2160001
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:1 mg
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:Abl kinase substrate
- Purity:95%
Specifications
About this item
This peptide, EAIYAAPFAKKK, was identified as a high affinity (Km=4 µM) and specific substrate for the Abl kinase through a degenerate peptide library.
The three lysines on the carboxy-terminus allow this peptide to be used in phosphocellulose kinase assays.