Src substrate
Catalog # ENZOBMLP1970001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:Src substrate
- Purity:95%
Specifications
About this item
This peptide, KVEKIGEGTYGVVYK, is derived from amino acids 6 to 20 of cdc2 and is a good substrate for src and other tyrosine kinases.