Order Entry
Belgium
ContactUsLinkComponent
IkappaB Kinase Substrat (Biotin)
IkappaB Kinase Substrat (Biotin)
Catalog # ENZOBMLP1480001
Supplier:  Enzo Life Sciences
CAS Number:  
IkappaB Kinase Substrat (Biotin)
Catalog # ENZOBMLP1480001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Biotin
  • Seq:
    02
  • Protein/peptide name:
    IkappaB kinase Substrate

Specifications

About this item

This peptide sequence, RHDSGLDSMKD, is derived from the area surrounding serine 32 and serine 36 of IκBα, which is phosphorylated by IκB Kinase (IKK).

The biotin allows the peptide to be used in IKK kinase assays with streptavidin-bound membranes.