Order Entry
Belgium
ContactUsLinkComponent
Fluor-de-Lys® H4-AcK16 deacetylase Substrate
Fluor-de-Lys® H4-AcK16 deacetylase Substrate
Catalog # ENZOBMLKI1740005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Fluor-de-Lys® H4-AcK16 deacetylase Substrate
Catalog # ENZOBMLKI1740005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Seq:
    02
  • Protein/peptide name:
    H4-AcK16 deacetylase substrate
  • Purity:
    95%
  • Formulation:
    Supplied as a 5 mM solution (100 µl) in HDAC assay buffer

Specifications

About this item

The deacetylated state of lysine-16 of histone H4 (H4-K16) is thought to be a critical factor in determining regions of chromatin silencing. Conversely, acetylation of lysine-16 can be associated with increased transcription, such as that due to dosage compensation from the X-chromosome of male Drosophila.

  • Acetylated H4 lysine-16 is preferentially deacetylated by yeast Sir2 in vitro and in vivo

Fluor de Lys®-H4-AcK16 is a fluorogenic, acetylated peptide substrate suitable for various lysine deacetylases, including the HDAC activities in HeLa nuclear extract, HDAC8, SIRT1 (human Sirtuin 1) and SIRT2 (human Sirtuin 2).

It is based on residues from histone H4 (12-16, Lys-Gly-Gly-Ala-Lys(Ac)) and was the preferred substrate for both SIRT1 and SIRT2 within a group patterned on acetylation sites in histone H3 and histone H4 N-terminal tails.

Fluor de Lys®-H4-AcK16 is deacetylated by SIRT1 (BML-SE239) at a rate of more than 6-fold and by SIRT2 (BML-SE251) at a rate of more than 40-fold those achieved with the acetylated lysine substrate, Fluor de Lys® (BML-KI104; acetylated substrates both at 25 µM, 500 µM NAD ).

Must be used in conjunction with Fluor de Lys® Developer (BML-KI104). Sufficient for 100 to 200 assays (50 to 100 µM substrate).