Order Entry
Belgium
ContactUsLinkComponent
Dendrotoxin-I (Dendroaspis p. polylepis)
Dendrotoxin-I (Dendroaspis p. polylepis)
Catalog # ENZOBMLKC1630140
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Dendrotoxin-I (Dendroaspis p. polylepis)
Catalog # ENZOBMLKC1630140
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    140 µG
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Protein/peptide name:
    Dendrotoxin-I (Dendroaspis p. polylepis)
  • Purity:
    >98% (HPLC)
  • Formulation:
    Lyophilised

Specifications

About this item

Ultra-pure

K⁺ channel blocker. Blocks Kv1.2 and Kv1.1 channels (IC50=0.13 and 3.1nM in oocytes respectively, with higher values for mammalian cells), as well as heteromultimeric channels comprised of Kv1.2 and Kv1.1 with other Kv1 isoforms.